missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SH3BP4 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877624
|
Proteintech
17691-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16867624
|
Proteintech
17691-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SH3BP4 was initially isolated as a novel cDNA using differential display analysis using cultured human corneal fibroblasts. The protein contains three Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. SH3BP4 has been shown to interact with specific endocytic proteins such as clathrin, dynamin, and the transferrin receptor (TfR) and localizes to TfR-containing coated pits and vesicles. It is thought to be involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of TfR. At least two isoforms of SH3BP4 are known to exist.Specifications
SH3BP4 | |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q921I6, Q9JJS5, Q9P0V3 | |
23677, 64634, 98402 | |
SH3BP4 Fusion Protein Ag11847 | |
Primary | |
-20°C | |
SH3BP4 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BOG25, EH binding protein 10, EHB10, SH3 domain binding protein 4, SH3BP4, TTP | |
Sh3bp4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title