Learn More
SH3BP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17691-1-AP-150UL
 View more versions of this product
Product Code. 16867624
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
SH3BP4 was initially isolated as a novel cDNA using differential display analysis using cultured human corneal fibroblasts. The protein contains three Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. SH3BP4 has been shown to interact with specific endocytic proteins such as clathrin, dynamin, and the transferrin receptor (TfR) and localizes to TfR-containing coated pits and vesicles. It is thought to be involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of TfR. At least two isoforms of SH3BP4 are known to exist.
Specifications
Specifications
SH3BP4
Polyclonal
Unconjugated
SH3BP4
BOG25, EH binding protein 10, EHB10, SH3 domain binding protein 4, SH3BP4, TTP
Rabbit
Antigen Affinity Chromatography
RUO
23677, 64634, 98402
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q921I6, Q9JJS5, Q9P0V3
Sh3bp4
SH3BP4 Fusion Protein Ag11847
150 μL
Primary
Human, Mouse, Rat