Learn More
Cardiac Troponin T Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Cardiac Troponin T |
---|---|
Concentration | 367 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16805015
|
Proteintech
26592-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16895005
|
Proteintech
26592-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cardiac Troponin T antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Cardiac Troponin T. This antibody recognizes human, mouse, rat antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cardiac Troponin T | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
367 μg/mL | |
Polyclonal | |
Rabbit | |
7139 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.