Learn More
Cardiac Troponin T Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26592-1-AP-150UL
Description
The Cardiac Troponin T antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Cardiac Troponin T. This antibody recognizes human, mouse, rat antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cardiac Troponin T | |
Polyclonal | |
Unconjugated | |
150 μL | |
7139 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.