Learn More
Cardiac Troponin T Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26592-1-AP-20UL
 View more versions of this product
Product Code. 16805015
  • 1645.54 SEK / 20µL
Description
The Cardiac Troponin T antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Cardiac Troponin T. This antibody recognizes human, mouse, rat antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Cardiac Troponin T
Polyclonal
Unconjugated
20 μL
7139
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
367 μg/mL
Rabbit
Primary
Human, Rat, Mouse