missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13899-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a subfamily of Rho GTPase activating proteins that contain a steroidogenic acute regulatory protein related lipid transfer domain. The encoded protein localizes to focal adhesions and may be involved in regulating cell morphology. This protein may also function as a tumor suppressor.Specifications
STARD8 | |
Polyclonal | |
Unconjugated | |
STARD8 | |
DLC 3, DLC3, KIAA0189, STARD8, START GAP3, STARTGAP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9754 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92502 | |
STARD8 | |
STARD8 Fusion Protein Ag4865 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction