Learn More
SIX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21305-1-AP-150UL
 View more versions of this product
Product Code. 16800915
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
SIX4 gene ontology annotations related to this gene include anatomical structure development; embryonic cranial skeleton morphogenesis; fungiform papilla morphogenesis; generation of neurons; inner ear morphogenesis; male gonad development; male sex determination; metanephric mesenchyme development; myoblast migration; myotome development; negative regulation of neuron apoptotic process; negative regulation of satellite cell differentiation; negative regulation of transcription, DNA-templated; olfactory placode formation; pharyngeal system development; positive regulation of branching involved in ureteric bud morphogenesis; positive regulation of ureteric bud formation; protein localization to nucleus; regulation of branch elongation involved in ureteric bud branching; regulation of epithelial cell proliferation; regulation of protein localization; regulation of synaptic growth at neuromuscular junction; sarcomere organization; skeletal muscle fiber differentiation; thymus development; trigeminal ganglion development.
Specifications
Specifications
SIX4
Polyclonal
Unconjugated
SIX4
AREC3, Homeobox protein SIX4, Sine oculis homeobox homolog 4, SIX homeobox 4, SIX4
Rabbit
Antigen Affinity Chromatography
RUO
20474, 299138, 51804
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q61321, Q9UIU6
SIX4
SIX4 Fusion Protein Ag15918
150 μL
Primary
Human, Mouse, Rat