missing translation for 'onlineSavingsMsg'
Learn More

P27/KIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen P27/KIP1
Concentration 0.77 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16805105
View Documents
Proteintech
26714-1-AP-20UL
20 μL
1645.54 SEK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16895095
View Documents
Proteintech
26714-1-AP-150UL
150 μL
4652.90 SEK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p27 Kip1 functions as a negative regulator of G1 mouse expression and has been proposed to function as a possible mediator of TGFb induced G1 arrest. p27 Kip1 is an important regulator of cell cycle progression, a potent inhibitor of cyclin E- and cyclin A-CDK2 complexes. p27 Kip1 forms a complex with cyclin type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. p27 Kip1 acts either as an inhibitor or an activator of cyclin type D-CDK4 complexes depending on its phosphorylation state and/or stoichiometry. p27 Kip1 has a molecular weight of approximately 27 kDa and the degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state.
Specifications

Specifications

P27/KIP1
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
P46414, P46527
1027, 12576
P27; KIP1 Fusion Protein Ag25083
Primary
-20°C
CDKN1B
0.77 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CDKN1B, CDKN4, KIP1, MEN1B, MEN4, P27, P27; KIP1, P27KIP1
CDKN1B
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.