missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH1-internal Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MYH1-internal |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16871825
|
Proteintech
22282-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16861825
|
Proteintech
22282-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscle contraction. Required for cytoskeleton organization.Specifications
MYH1-internal | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P12882 | |
4619 | |
Peptide | |
Primary | |
-20°C | |
MYH1 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MYH1, MYHa, MyHC 2x, MyHC 2X/D, MyHC IIx/d, MYHSA1, Myosin 1, Myosin heavy chain 1, Myosin heavy chain 2x, Myosin heavy chain IIx/d | |
MYH1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title