missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH1-internal Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22282-1-AP-150UL
This item is not returnable.
View return policy
Description
Muscle contraction. Required for cytoskeleton organization.Specifications
MYH1-internal | |
Polyclonal | |
Unconjugated | |
MYH1 | |
MYH1, MYHa, MyHC 2x, MyHC 2X/D, MyHC IIx/d, MYHSA1, Myosin 1, Myosin heavy chain 1, Myosin heavy chain 2x, Myosin heavy chain IIx/d | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4619 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12882 | |
MYH1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction