Learn More
Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
CAS: 4039-32-1 | C6H18LiNSi2 | 167.327 g/mol
1015.00 SEK - 3965.00 SEK
Chemical Identifiers
CAS | 4039-32-1 |
---|---|
Molecular Formula | C6H18LiNSi2 |
Molecular Weight (g/mol) | 167.327 |
MDL Number | MFCD00008261 |
InChI Key | YNESATAKKCNGOF-UHFFFAOYSA-N |
Synonym | lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide |
PubChem CID | 2733832 |
IUPAC Name | lithium;bis(trimethylsilyl)azanide |
SMILES | [Li+].C[Si](C)(C)[N-][Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324857
|
Thermo Scientific Alfa Aesar
041872.22 |
100 g |
1015.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314857
|
Thermo Scientific Alfa Aesar
041872.36 |
500 g |
3965.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium bis(trimetylsilyl)amide is used as a strong base in organic synthesis and as catalysts in polymerization reactions. It is also used to form various organolithium compounds such as acetylides and lithium enolates. It is a base employed in generating enolates for preparation of lactone precursors, pyranones and cyclohexanes. Further, it is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. In addition, it is used in a three-step synthesis of disubstituted 1,2,5-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
4039-32-1 | |
167.327 | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
2733832 | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C |
C6H18LiNSi2 | |
MFCD00008261 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
lithium;bis(trimethylsilyl)azanide |
Specifications
4039-32-1 | |
0.716 g/mL | |
ChemSeal™ bottles | |
[(CH3)3Si]2NLi | |
UN2924 | |
Air and moisture sensitive | |
Soluble in tetrahydrofuran,hexane and toluene. | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.327 | |
167.33 | |
Lithium bis(trimethylsilyl)amide |
Colorless to Yellow | |
-31°C (-24°F) | |
C6H18LiNSi2 | |
MFCD00008261 | |
8136861 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H304-H314-H335-H336-H361f-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen