Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data
For one free-selectable temperature value
19145.00 SEK
Specifications
Product Type IQ/OQ Document with Device-specific Works Test Data
View More Specs
Products 6
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17130305
View Documents
Memmert™
WTB11D00125
WTB11
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
17130335
View Documents
Memmert™
WTB15D00125
WTB15
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
17130365
View Documents
Memmert™
WTB24D00125
WTB24
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
17140425
View Documents
Memmert™
WTB6D00125
WTB6
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
17160385
View Documents
Memmert™
WTB35D00125
WTB35
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
17190405
View Documents
Memmert™
WTB50D00125
WTB50
19145.00 SEK
Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Includes temperature distribution survey at Memmert
  • For 9 measuring points to DIN 12876.2
Specifications
Specifications
IQ/OQ Document with Device-specific Works Test Data