Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data
For one free-selectable temperature value
Brand:  Memmert™ WTB15D00125
 View more versions of this product
Product Code. 17130335
  • 19145.00 SEK / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Includes temperature distribution survey at Memmert
  • For 9 measuring points to DIN 12876.2
Specifications
Specifications
WTB15
IQ/OQ Document with Device-specific Works Test Data