Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
For use with Fisherbrand vortex mixers.
703.00 SEK - 1155.00 SEK
Specifications
Material | Foam |
---|---|
For Use With (Equipment) | Digital and analog mini vortex mixer |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15736949
|
Fisherbrand™
80862997 |
Microtube Foam rack |
703.00 SEK
Pack of 2 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
15746949
|
Fisherbrand™
80862998 |
Microplate Holder |
1155.00 SEK
Pack of 2 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
- Mixes 96-well plates or deepwell blocks
- Requires Insert Retainer (02-215-391)
Specifications
Foam | |
Digital and analog mini vortex mixer |