Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80862997
483.42 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15736949
  • 703.00 SEK / Pack of 2
Only null left
Estimated Shipment: 14-08-2025
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Mixes 96-well plates or deepwell blocks
  • Requires Insert Retainer (02-215-391)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.5 and 0.5 mL
Microtube Foam rack
Foam
Digital and analog mini vortex mixer