Learn More
3-Methyl-2-phenoxybutyryl chloride, 98%, Thermo Scientific™
1083.00 SEK - 4851.00 SEK
Chemical Identifiers
CAS | 65118-10-7 |
---|---|
Molecular Formula | C11H13ClO2 |
Molecular Weight (g/mol) | 212.673 |
MDL Number | MFCD02262198 |
InChI Key | WOKLTCPOXPCYSM-UHFFFAOYSA-N |
Synonym | iso-propylphenoxyacetyl chloride, 3-methyl-2-phenoxybutyryl chloride, acmc-1bag3, butanoyl chloride,3-methyl-2-phenoxy, 3-methyl-2-phenoxybutanoic acid chloride |
PubChem CID | 11298820 |
IUPAC Name | 3-methyl-2-phenoxybutanoyl chloride |
SMILES | CC(C)C(C(=O)Cl)OC1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesMoisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
Chemical Identifiers
65118-10-7 | |
212.673 | |
WOKLTCPOXPCYSM-UHFFFAOYSA-N | |
11298820 | |
CC(C)C(C(=O)Cl)OC1=CC=CC=C1 |
C11H13ClO2 | |
MFCD02262198 | |
iso-propylphenoxyacetyl chloride, 3-methyl-2-phenoxybutyryl chloride, acmc-1bag3, butanoyl chloride,3-methyl-2-phenoxy, 3-methyl-2-phenoxybutanoic acid chloride | |
3-methyl-2-phenoxybutanoyl chloride |
Specifications
65118-10-7 | |
MFCD02262198 | |
Moisture sensitive | |
WOKLTCPOXPCYSM-UHFFFAOYSA-N | |
3-methyl-2-phenoxybutanoyl chloride | |
11298820 | |
98% |
C11H13ClO2 | |
UN3265 | |
iso-propylphenoxyacetyl chloride, 3-methyl-2-phenoxybutyryl chloride, acmc-1bag3, butanoyl chloride,3-methyl-2-phenoxy, 3-methyl-2-phenoxybutanoic acid chloride | |
CC(C)C(C(=O)Cl)OC1=CC=CC=C1 | |
212.673 | |
212.68 | |
3-Methyl-2-phenoxybutyryl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures