Learn More
3-Methyl-2-phenoxybutyryl chloride, 98%, Thermo Scientific™
Product Code. 11375429
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65118-10-7
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD02262198
InChI Key WOKLTCPOXPCYSM-UHFFFAOYSA-N
Synonym iso-propylphenoxyacetyl chloride, 3-methyl-2-phenoxybutyryl chloride, acmc-1bag3, butanoyl chloride,3-methyl-2-phenoxy, 3-methyl-2-phenoxybutanoic acid chloride
PubChem CID 11298820
IUPAC Name 3-methyl-2-phenoxybutanoyl chloride
SMILES CC(C)C(C(=O)Cl)OC1=CC=CC=C1
Specifications
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 212.68
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-phenoxybutyryl chloride