Learn More
Zinc Standard Solution Concentrate, 100ppm, Reagecon™
BIOPHARMA
Product Code. 11404779 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Packaging 100mL
Concentration 100 ppm
Quantity 100 mL
Product Identifier
  • CONCENTRATE TO MAKE (5003401)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : zinc sulphate (hydrous) (mono-, hexa-andhepta hydrate)