missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, ReagentPlus™, 99.9%, Honeywell™

Product Code. 15671760 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, powder, <5 μm particle size, 99.9%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Color White
Physical Form Powder
Quantity 1 kg
Linear Formula ZnO
UN Number UN3077
Formula Weight 81.39g/mol
Percent Purity 99.90%
Grade ReagentPlus™
Packaging Poly Bottle
Density 5.6g/cu.cm
Chemical Name or Material Zinc Oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.