missing translation for 'onlineSavingsMsg'
Learn More

Zinc-alpha-2-glycoprotein Mouse anti-Human, Clone: 4B8A4, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen Zinc-alpha-2-glycoprotein
Clone 4B8A4
Concentration 0.98 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16867815
View Documents
Proteintech
66178-1-IG-20UL
20 μL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16857815
View Documents
Proteintech
66178-1-IG-150UL
150 μL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.
Specifications

Specifications

Zinc-alpha-2-glycoprotein
0.98 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AZGP1, ZA2G, ZAG, Zinc alpha 2 glycoprotein, Zinc-alpha-2-glycoprotein, Zn alpha 2 glycoprotein, Zn alpha 2 GP, ZNGP1
AZGP1
IgG1
Protein G
Antibody
4B8A4
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
P25311
563
Zinc-alpha-2-glycoprotein Fusion Protein Ag5661
Primary
-20°C
AZGP1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.