missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YWHAZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14881-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was raised against the full-length YWHAZ.
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathways. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Several transcript variants that differ in the 5′ UTR but that encode the same protein have been identified for this gene.Specifications
YWHAZ | |
Polyclonal | |
Unconjugated | |
YWHAZ | |
14 3 3 protein zeta/delta, 14-3-3, KCIP 1, YWHAZ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22631, 25578, 7534 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63101, P63102, P63104 | |
YWHAZ | |
YWHAZ Fusion Protein Ag6654 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction