Learn More
Berkshire™ 360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Brand: Berkshire™ EC360.EC.6
6056.68 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
EasyClean 360 ICT Non-Sterile Knitted Polyester Elastic and Padded Covers (17-987-361)
- Made from Berkshire's SuperPolx™ 1200A 100% continuous filament knitted polyester material
- Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
- Non-shedding and low-linting cover per USP requirements
- Stretch fabric and elastic edge for easy on/off with secure fit on mop head
- Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
- Compatible for use in ISO Class 3/EU Grade A Environments after sterilizing
- Non-sterile
Specifications
Isolator Cleaning Tool Cover | |
Non-sterile | |
6/Pk. | |
EasyClean™ 360 Isolator Cleaning Tool | |
Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments. | |
Knitted Polyester Elastic |
White | |
Autoclavable | |
25 knitted polyester elastic covers and 1 knitted polyester padded cover/pack | |
Mop Covers | |
ISO Class 3/EU Grade A |