Learn More
Kimberly-Clark™ WypAll™ L40 Power Clean™ Cleaning Wipers
Wipers deliver softness and strength, perfect for routine cleaning or for wiping hands and faces. Strong, very thick and highly absorbent. Disposable white cloths made with DRC base sheet technology clean up fast without falling apart.
3005.00 SEK - 3560.00 SEK
Specifications
Product Type | Medium-duty Industrial Wiper |
---|---|
Material | Double Re-crepe Cellulose |
Color | White |
For Use With (Application) | A limited life wiper using double re-creped (DRC) base sheet technology, layered fabric that’s strong, thick and highly absorbent so it cleans up fast without falling apart, does the job with fewer wipers and helps reduce costs |
Ply | 1-ply |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
11778038
![]() |
Kimberly-Clark™
7462 |
25 x 41 cm (9.8 x 16.1 in.) |
3560.00 SEK
Pack of 9 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
10627645
![]() |
Kimberly-Clark™
7471 |
30.4 x 31.7 cm (12 x 12.5 in.) |
3005.00 SEK
Pack of 18 |
In Stock
Log in to see stock. |
|||||
Description
- Exceptional absorption that is 25% faster than regular towels
- Made with Double Re-Creped (DRC) base sheet technology, layered fabric that is strong, thick, and highly absorbent
- Compatible for use in spray and wipe applications with peracetic acid, hydrogen peroxide, bleach (sodium hypochlorite) and quats (quaternary ammonium compounds)
- Always follow manufacturers' instructions for dilution, application, and contact time
- Flexible system to give you controlled access to hygienically protected wipers when cleaning or wiping on the move
Specifications
Medium-duty Industrial Wiper | |
White | |
1-ply |
Double Re-crepe Cellulose | |
A limited life wiper using double re-creped (DRC) base sheet technology, layered fabric that’s strong, thick and highly absorbent so it cleans up fast without falling apart, does the job with fewer wipers and helps reduce costs | |
Non-sterile |