Learn More
3M™ White with Red Seams Polypropylene/Polyethylene Laminate Film Protective Coveralls
2501.04 SEK
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Disposable | Single-use |
Waist Style | Elastic |
Cuff Style | Knit |
Description
Specifications
General Purpose Coveralls | |
Single-use | |
Knit | |
3-panel | |
Category III |
Unisex | |
Elastic | |
Hooded | |
Non-sterile |