Learn More
DWK Life Sciences Wheaton™ Safety-Coated Narrow Mouth Amber Bottles with Aluminum-Foil-Lined Black Phenolic Caps
Retain contents and most glass fragments for safer and easier disposal
2005.00 SEK - 3325.00 SEK
Specifications
Coating | Plastisol |
---|---|
Color | Amber |
Material | Plastisol-coated Soda Lime Glass |
Closure Color | Black |
Certifications/Compliance | USP Type III |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
12442561
|
DWK Life Sciences
220935 |
500 mL | 28-400 mm |
3325.00 SEK
Pack of 24 |
Estimated Shipment: 01-12-2025
Log in to see stock. |
||||
12452561
|
DWK Life Sciences
220936 |
1000 mL | 33-400 mm |
2005.00 SEK
Pack of 12 |
Estimated Shipment: 01-12-2025
Log in to see stock. |
||||
Description
- Plastisol-coated soda-lime glass
- Coating is inert and highly resistant to strong acids, alkalis, and most solvents
- Coating reduces risk of personal injury or contamination
- Black, foil-lined caps included
- Not autoclavable
Specifications
Plastisol | |
Plastisol-coated Soda Lime Glass | |
USP Type III | |
Aluminum Foil | |
Screw Cap |
Amber | |
Black | |
Yes | |
Phenolic |