Learn More
Cytiva Whatman™ GV 025 Series Glass Vacuum Filtration Manifolds, Replacement Parts
Replacement Parts for Whatman™ GV 025 Series Vacuum Filtration Manifolds. Includes flasks, supports, funnels, frits, centering rings, and clamps.
346.00 SEK - 1820.00 SEK
Product Code | Brand | Type | Model | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Model | For Use With (Equipment) | Price | Quantity & Availability | |||
17819991
|
Cytiva
10441002 |
Manifold Base | GV 025/0/02 | Manifold Model GV 025/0 (11349274) |
685.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||
11314924
|
Cytiva
10441203 |
Erlenmeyer Flask | GV 025/2/08 | Manifold Model GV 025/2 |
1360.00 SEK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||
11354914
|
Cytiva
10441001 |
Funnel | GV 025/0/01 | Manifold Model GV 025/0 |
1005.00 SEK
Each |
In Stock
Log in to see stock. |
|||
11304924
|
Cytiva
10441201 |
Manifold Base | GV 025/2/02 | Manifold Model GV 025/2 |
1820.00 SEK
Each |
In Stock
Log in to see stock. |
|||
11924556
|
Cytiva
10441004 |
Centering Ring | GV 025/0/05 | Manifold Model GV 025/0 |
346.00 SEK
Each |
In Stock
Log in to see stock. |
|||
11374914
|
Cytiva
10441003 |
Frit | GV 025/0/03 | Manifold Model GV 025/0 |
645.00 SEK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||
11394914
|
Cytiva
10441005 |
Spring Action Clamp | GV 025/0/06 | Manifold Model GV 025/0 | N/A | ||||
Description
- Description: Replacement Part for GV 025 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Vacuum Filtration |