Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
Brand:  Kartell™ 0104300
 View more versions of this product
Product Code. 15845497
  • 487.00 SEK / Pack of 10
Only null left
Estimated Shipment: 18-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Designed to prevent machined glass joints from binding
  • Conforms to DIN 12242
  • Suitable for foodstuff
Specifications
Specifications
DIN 12242
45 mm
Prevent glass joints from binding
45/40 Cone Size
PTFE
Glass Joint
10/Pk.