Learn More
Brady™ BradyGlo™ Photoluminescent Polyester Glow-in-the-dark Arrow Tape
BradyGlo™ B-324 arrow tapes exceed ASTM E 2072-00 standard specifications as well as the standards for 10+ hour glow requirements.
Brand: Brady™ 090973
1632.26 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- BradyGlo™ B-324 arrow tapes exceed ASTM E 2072-00 standard specifications as well as the following standards for 10+ hour glow requirements: ASTM/E162/E648/E662, SMP800C, IMO Resolution A. 752(18), ISO/CD 15370, PSPA Standard 002 Part 2 1993 Class A Rev. 2: 09/99, DIN 67 510, Parts 1-4, NFPA 101 Life Safety Code 2000 7.10.7.2, OSHA 1910.37 and Lloyd's register Cert. 01/6007
Specifications
ASTM E–2072-00 / ASTM E–2073-00, ASTM E-162, ASTM E-648, ASTM E-662, IMO Resolution A.752(18), ISO/CD 15370, Marine Safety Committee MSC.27(61), DIN 67 510 (Parts 1 - 4), and PSPA Standard 002 Part 2 | |
Permanent Acrylic, Pressure-sensitive | |
Polyester | |
For exit and directional signs, identification of fire alarms and extinguishers, low location egress pathway markings, and directional systems for escape or evacuations routes. Recommended Uses of Photoluminescent Safety Markings | |
-40°C | |
1 Ea. | |
-40°F to +176°F | |
0.0079 in. | |
4.57 m x 50.80 mm | |
4.57 m | |
50.8 mm | |
B324 2 in. X5YD GRN/GLO ARROW TAPE |
Arrow-Style | |
Green on Photoluminescent | |
Photoluminescent Tape | |
60° 121 Gardner Units | |
1 Roll | |
Arrow | |
-40°C to +80°C | |
0.201 mm | |
5 yd. | |
2 in. | |
2 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction