Learn More
Chroma Gesellschaft™ Closed Stand Rings with Bosshead
Provides for ease of connection on ring stands. Chroma Gesellschaft™ Closed Stand Rings with Bosshead are manufactured from steel.
Brand:  Chroma Gesellschaft™ 2013/2
Product Code. 11709037
  • 208.00 SEK / Each
Only null left
Estimated Shipment: 08-08-2025
Add to basket
Description
Description
  • Bosshead with plastic-headed clamping screw
  • Accepts rods from 8mm to 12.5mm in diameter
  • Dimensions given are internal diameter of ring x center to center distance from bosshead to ring
Specifications
Specifications
50mm Bosshead
Closed Ring
1 Ea.