Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand: SCAT™ 107993
798.72 SEK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
GL32 (f) to GL45 (m) | |
PTFE | |
Thread Adapter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.