Learn More
Versa Rx Checkweigher for Pharmaceutical Applications
Meet the high performance standards of the pharmaceutical industry with the Thermo Scientific™ Versa Rx Checkweigher.
Brand: Thermo Scientific VERSARXPHARMA
Description
Meet the high performance standards of the pharmaceutical industry with the Thermo Scientific™ Versa Rx Pharmaceutical Checkweigher, which provide trusted reliability so you can protect your brand in the pharmaceutical (or food) industry.
With high-accuracy—within ±50mg at 3 sigma and speeds up to 650ppm—the Versa Rx Pharma checkweigher also offers:
- Easy use, easy maintenance and sanitary design
- Slack belt knife edge conveyor design for unparalleled accuracy and optimal product handling
- Ideal for a variety of products (blister packs, cartons, plastic bottles, glass bottles, pouches and small aerosols)
- 21 CFR Part 11 compliance option
- Fail-safe reject mechanisms and reject validation option
- OIML MID approved
Recommended for: Pharmaceutical and food industries
Specifications
5.5 bar (80 psi) 600 kPa | |
USB; serial; parallel; FTP Ethernet; Modbus | |
Up to 650 ppm | |
Yes | |
115/230 VAC 50/60 Hz | |
50/60 Hz | |
Stainless Steel | |
15 amps | |
115/230 VAC | |
Slack belt: 2 - 300 g Taut belt: 10 - 500 g | |
±50 mg at 3 sigma |
CE | |
Versa Rx Pharmaceutical Checkweigher | |
Air Jet, Air Bopper, Air Flipper | |
No | |
24.6 to 39.4 in. | |
625 to 1000 mm | |
5°C to 40°C (41°F to 104°F) | |
20 - 50% non-condensing | |
Slack belt: 0.07 - 10.57 oz. Taut belt: 0.35 - 17.62 oz. | |
Versa Rx Pharmaceutical Checkweigher |