Learn More
DuPont™ Tyvek 500 Trousers
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ 500 Trousers offer an excellent barrier against fine particles and fibers down to 1 micron in size.
Brand:  DuPont™ D13496109
 View more versions of this product
Product Code. 12365659
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Flash-spun HDPE construction
  • Elasticated waist
  • Stitched seams
  • No pockets
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Medium
Flashspun HDPE
CE Certified, EN 1149-5
Applications includes pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Trousers
White
Elastic
Internal Stitched Inseam