Learn More
DuPont™ Tychem 6000 F Laboratory Gowns
Shop All DuPont Products
Click to view available options
Size: Small to Medium
Description
- Elasticated wrists
- Antistatic treatment
- With wrap-over rear closure, hook and loop neck closure and waist ties
- Excellent chemical permeation protection to an extensive range of chemicals
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Specifications
Size | Small/Medium |
Gender | Unisex |
No. of Pockets | None |
Style | Shin Length |
Product Type | General Purpose Gown |
Material | Tychem |
Color | Gray |
Closure Type | Wrap-around Waist Ties |
Disposable | Single-use |
Sterility | Non-sterile |