Learn More
DuPont™ Tychem™ 6000 F FaceSeal
Tychem™ 6000 F FaceSeal is a hooded coverall that includes a rubber seal for a tight fit.
1595.00 SEK - 1800.00 SEK
Specifications
Color | Gray |
---|---|
Ankle Style | Attached socks |
Closure Type | Rear-entry zipper with double flap, snaps and hook-and-loop seal |
Cuff Style | Attached gloves |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16328555
|
DuPont™
D15490341 |
Small |
1595.00 SEK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
16348555
|
DuPont™
D15490481 |
Medium |
1595.00 SEK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
16338555
|
DuPont™
D15490467 |
Large |
1595.00 SEK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
16358555
|
DuPont™
D15490598 |
2X-Large |
1595.00 SEK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
16368555
|
DuPont™
D15490650 |
3X-Large |
1675.00 SEK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
16378555
|
DuPont™
D15490706 |
4X-Large |
1735.00 SEK
Each |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
16388555
|
DuPont™
D15490762 |
5X-Large |
1800.00 SEK
Each |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
Description
- Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
- Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) on inside
- Stitched and over-taped seams with barrier tape for protection and strength
- Back entry with zipper, double zipper flap with snaps and hook and loop closure
- Attached undergloves
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Gray | |
Rear-entry zipper with double flap, snaps and hook-and-loop seal | |
Single-use | |
Hooded | |
Stitched, Taped | |
Elastic |
Attached socks | |
Attached gloves | |
Unisex | |
1/Ea. | |
Non-sterile |