Learn More
Kinesis™ Tubing PTFE 20m 1/16inch O.D, 0.30mm I.D. Omnifit
Brand:  Kinesis™ 008T16-030-20
Encompass_Preferred
Product Code. 12916291
  • POA / 20m
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.