Learn More
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
Description
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | >300°C |
Beilstein | 2207452 |
Merck Index | 14,9569 |
Solubility Information | Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol. |
Quantity | 250 g |
Formula Weight | 157.21 |
Percent Purity | 97% |
Chemical Name or Material | trans-4-(Aminomethyl)cyclohexanecarboxylic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.