Learn More
TPI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10713-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.Specifications
TPI1 | |
Polyclonal | |
Unconjugated | |
TPI1 | |
TIM, TPI, TPI1, Triose phosphate isomerase, Triosephosphate isomerase, triosephosphate isomerase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21991, 24849, 7167 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17751, P48500, P60174 | |
TPI1 | |
TPI1 Fusion Protein Ag1175 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |