Learn More
TNF Alpha Mouse anti-Human, CoraLite 488, Clone: 7B8A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60291-100UL
Description
The TNF Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNF Alpha. This antibody recognizes human antigen. The TNF Alpha antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
TNF Alpha | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
7124 | |
IgG2b |
Western Blot, Immunofluorescence | |
7B8A11 | |
Mouse | |
Primary | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.