Learn More
TNF Alpha Mouse anti-Human, CoraLite 488, Clone: 7B8A11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60291-100UL
Product Code. 16880616
  • 5106.85 SEK / 100µL
Description
The TNF Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNF Alpha. This antibody recognizes human antigen. The TNF Alpha antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
TNF Alpha
Monoclonal
CoraLite 488
100 μL
7124
IgG2b
Western Blot, Immunofluorescence
7B8A11