Learn More
IKA™ Tip XL Box
Box of 10 mL pipette tips that include calibration rings.
Brand:  IKA™ 0020017832
1648.25 SEK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Product Code. 16248541
  • 1735.00 SEK / Pack of 250
Genomics Powerhouse
Use promo code 24117 at checkout
View Available Promotions
Only null left
In Stock
Description
  • Includes 10 stackpacks
  • Transparent
  • With calibration rings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL
Calibration Rings
Pipette Tips
Transparent
25/Box, 10 Boxes/Pk
Tip XL