Learn More
Tin(Ii) Chloride Dihydrate, Reagent grade, Honeywell™
Product Code. 15651810 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
Reagent Grade, 98%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10025-69-1
MDL Number MFCD00149863
Specifications
Quantity 250 g
Linear Formula SnCl2 · 2H2O
UN Number 3260
Solubility Information Soluble in water
Formula Weight 225.65g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material Tin(Ii) Chloride Dihydrate