Learn More
Tetraethylsilane, 97%, Thermo Scientific Chemicals
Product Code. 11332106
Click to view available options
Quantity:
5 g
25 g
100 g
CAS: 631-36-7 | C8H20Si | 144.333 g/mol
Used as the intermediates of OLED, pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as the intermediates of OLED, pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.333
MDL Number MFCD00009019
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC
Specifications
Boiling Point 153°C to 155°C
Melting Point -82°C
Quantity 25 g
UN Number UN1993
Beilstein 1732768
Flash Point 26°C (78°F)
Solubility Information Insoluble in water.
Formula Weight 144.33
Percent Purity 97%
Odor Recognizable
Refractive Index 1.426
Density 0.767
Chemical Name or Material Tetraethylsilane