Learn More
Tetrabutylammonium Hydroxide Solution, 0.1M, Honeywell Fluka™
Product Code. 15617360 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
1 L
Volumetric, 0.1 M (C4H9)4NOH (isopropanol/methanol (10:1 V/V)), for titration in non-aqueous medium
Honeywell´s high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
Specifications
Flash Point 15°C (59°F)
Linear Formula (CH3CH2CH2CH2)4N(OH)
Quantity 1 L
UN Number UN1992
Beilstein 3571228
IUPAC Name tetrabutylazanium hydroxide
Formula Weight 259.47g/mol
Chemical Name or Material Tetrabutylammonium hydroxide solution