Learn More
Hamilton™ Terasaki Dispenser
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
Brand:  Hamilton™ 201920
12188.34 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10008330
  • 12495.00 SEK / Each
Description
  • Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
  • Uses removable needles
  • Available for a range of volumes
Specifications
Specifications
1/Pk.
Terasaki Dispenser
100 μL