Learn More
Isofield™ Tera 230 Validated Sterile Tie-On Cleanroom Facemask - Grade A/B Compatible
Click to view available options
:
Size: Wide
Description
- Validated sterile tie-on cleanroom facemask
- 230 mm length with malleable nose strip
- 3-layer non-woven configuration
- Highly efficient bacterial and particle filtration
- Gamma irradiation, minimum 25 kGy
- Sterility Assurance Level 10-6
- Particle Shedding (Helmke Drum) Category I Individually packed pouches printed with IPA resistant ink to maintain traceability
Specifications
Specifications
Product Type | Cleanroom Face Mask |
Color | White |
Fastening Type | Ties |
Material | Polyethylene, Polypropylene |
Size | Wide |
Sterility | Sterile |
Style | Pleated |