Learn More
Memmert™ Temperature Restriction for UF55
Optional temperature restriction between 60°C and 250°C.
Brand:  Memmert™ UF55A8
1815.77 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17549425
  • 1900.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Accessory
  • 1 Choice
  • Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
Specifications
UF55 Oven
Temperature Restriction