missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 95–97%, Reagent Grade, Scharlab
Click to view available options
Quantity:
1 L
2.5 L
Description
- Molecular Formula: H2SO4
- Molar Mass: 98.08g/mol
- CAS: 7664-93-9li>
- EIN ECS No.: 231-639-5
Specifications
Specifications
Melting Point | ∼-15°C |
Boiling Point | ∼310°C |
Density | 1.84g/cm3 |
Residue after Ignition | 0.0003% max |
Vapor Pressure | ∼0.0001hPa at 20°C |
Quantity | 2.5 L |
Physical Form | Solution |
Solubility Information | Miscible in water (20°C) |
Formula Weight | 98.08g/mol |
Percent Purity | 95 to 97% |
Show More |
Safety and Handling

Product Identifier
- Sulfuric acid, 95 -97 %, reagent grade, ISO
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Sulphuric Acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction