Learn More
Memmert™ Subframe for UF55, Adjustable up to 600 mm
Adjustable subframe for Memmert Ovens.
Brand:  Memmert™ UF55B29747
3847.16 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17549435
  • 4020.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Adjustable in height
Specifications
Specifications
Adjustable
UF55 Oven
Subframe
Up to 600 mm