Learn More
Memmert™ Subframe for UF55, Adjustable up to 130 mm
Adjustable subframe for Memmert Ovens.
Brand: Memmert™ UF55B33659
263.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adjustable in height
Specifications
Adjustable | |
UF55 Oven | |
Subframe | |
Up to 130 mm |