Learn More
Memmert™ Subframe on Castors for UF55
Subframe on wheels for Memmert Ovens.
Brand: Memmert™ UF55B29748
5696.97 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- On Castors
- Height: 660 mm
Specifications
On Wheels | |
UF55 Oven | |
Subframe | |
660 mm |