Learn More
Sampling Systems™ Sub Sampler Pro
Can sample from a set distance from the bottom of a container or alternatively at a mid point in a container or well. Sampling Systems™ Sub Sampler Pro is sometimes called a Bomb Sampler and it is used throughout the world for both chemical and environmental sampling.
3730.00 SEK - 11390.00 SEK
Specifications
Product Type Sampler
Capacity (Metric) 500 mL
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
15809361
View Documents
Sampling Systems™
5072B-500
Stainless Steel
3730.00 SEK
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 12-09-2025
Add to basket
15899351
View Documents
Sampling Systems™
5072A-500
Nickel-plated Brass
11390.00 SEK
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 12-09-2025
Add to basket
Description
Description
  • Lower Sub Sampler Pro until the plunger assembly reaches the bottom of the container
  • When the plunger assembly hits the bottom, it is pushed up allowing the sample to enter the device
  • When the sampler is raised the plunger will drop down and seal the sampler
  • Lower the Sub Sampler Pro into the liquid
  • At the required depth pull on the control line to raise the plunger
  • This will allow the liquid to enter the sampler
  • Release the control Line and the plunger will drop down and seal the sample
Specifications
Specifications
Sampler
500 mL